MilliporeSigma
On the concentration-induced growth of nonionic wormlike micelles.
The European physical journal. E, Soft matter (2008-10-31)
S Bulut, J Hamit, U Olsson, T Kato
ABSTRACT
We present a comprehensive study of the concentration-induced growth of nonionic wormlike micelles in dilute solutions, below the overlap concentration, where we combine static and dynamic light scattering and NMR self-diffusion. The data are analyzed in detail in terms of the number-averaged contour length, assuming an exponential size distribution, as predicted by theory, and that the micellar flexibility can be described by the wormlike chain model with a certain persistence length. A very good agreement between the different experimental methods is obtained. The number-averaged contour length increases with increasing micelle volume fraction. The data are consistent with a power law, where the power law exponent is in the range 0.5-0.8, depending on the system. The result is in reasonable agreement with the theory of living polymers.
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)