MilliporeSigma
  • Fluorescence probing of aminofluorene-induced conformational heterogeneity in DNA duplexes.
Fluorescence probing of aminofluorene-induced conformational heterogeneity in DNA duplexes.
Chemical research in toxicology (2008-01-16)
Nidhi Jain, Yana K Reshetnyak, Lan Gao, M Paul Chiarelli, Bongsup P Cho
ABSTRACT
Fluorescence spectroscopy was used to study carcinogen-induced conformational heterogeneity in DNA duplexes. The fluorophore 2-aminopurine (AP) was incorporated adjacent (5') to the lesion (G*) in eight different DNA duplexes [d(5'-CTTCT PG* NCCTC-3'):d(5'-GAGGN XTAGAAG-3'), G* = FAF adduct, P = AP, N = G, A, C, T, and X = C, A] modified by FAF [ N-(2'-deoxyguanosin-8-yl)-7-fluoro-2-aminofluorene], a fluorine-tagged model DNA adduct derived from the potent carcinogen 2-aminofluorene. Steady-state measurements showed that fluorescence intensity and Stern-Volmer constants ( Ksv) derived from acrylamide quenching experiments decreased for all carcinogen-modified duplexes relative to the controls, which suggests greater AP stacking in the duplex upon adduct formation. Conformation-specific stacking of AP with the neighboring adduct was evidenced by a sequence-dependent variation in fluorescence intensity, position of emission maximum, degree of emission quenching by acrylamide, and temperature-dependent spectral changes. The magnitude of stacking was in the order of FAF residue in base-displaced stacked (S) > minor groove wedged (W) > major groove B type (B). This work represents a novel utility of AP in probing adduct-induced conformational heterogeneities in DNA duplexes.
Sigma-Aldrich
2-Aminofluorene, 98%