MilliporeSigma
  • Metabolic response to an acute jump in cardiac workload: effects on malonyl-CoA, mechanical efficiency, and fatty acid oxidation.
Metabolic response to an acute jump in cardiac workload: effects on malonyl-CoA, mechanical efficiency, and fatty acid oxidation.
American journal of physiology. Heart and circulatory physiology (2007-12-18)
Lufang Zhou, Hazel Huang, Celvie L Yuan, Wendy Keung, Gary D Lopaschuk, William C Stanley
ABSTRACT
Inhibition of myocardial fatty acid oxidation can improve left ventricular (LV) mechanical efficiency by increasing LV power for a given rate of myocardial energy expenditure. This phenomenon has not been assessed at high workloads in nonischemic myocardium; therefore, we subjected in vivo pig hearts to a high workload for 5 min and assessed whether blocking mitochondrial fatty acid oxidation with the carnitine palmitoyltransferase-I inhibitor oxfenicine would improve LV mechanical efficiency. In addition, the cardiac content of malonyl-CoA (an endogenous inhibitor of carnitine palmitoyltransferase-I) and activity of acetyl-CoA carboxylase (which synthesizes malonyl-CoA) were assessed. Increased workload was induced by aortic constriction and dobutamine infusion, and LV efficiency was calculated from the LV pressure-volume loop and LV energy expenditure. In untreated pigs, the increase in LV power resulted in a 2.5-fold increase in fatty acid oxidation and cardiac malonyl-CoA content but did not affect the activation state of acetyl-CoA carboxylase. The activation state of the acetyl-CoA carboxylase inhibitory kinase AMP-activated protein kinase decreased by 40% with increased cardiac workload. Pretreatment with oxfenicine inhibited fatty acid oxidation by 75% and had no effect on cardiac energy expenditure but significantly increased LV power and LV efficiency (37 +/- 5% vs. 26 +/- 5%, P < 0.05) at high workload. In conclusion, 1) myocardial fatty acid oxidation increases with a short-term increase in cardiac workload, despite an increase in malonyl-CoA concentration, and 2) inhibition of fatty acid oxidation improves LV mechanical efficiency by increasing LV power without affecting cardiac energy expenditure.
Sigma-Aldrich
4-Hydroxy-L-phenylglycine, ≥99.0% (NT)