MilliporeSigma
  • Chiral salen-aluminum complex as a catalyst for enantioselective alpha-addition of isocyanides to aldehydes: asymmetric synthesis of 2-(1-hydroxyalkyl)-5-aminooxazoles.
Chiral salen-aluminum complex as a catalyst for enantioselective alpha-addition of isocyanides to aldehydes: asymmetric synthesis of 2-(1-hydroxyalkyl)-5-aminooxazoles.
Organic letters (2007-08-10)
Shi-Xin Wang, Mei-Xiang Wang, De-Xian Wang, Jieping Zhu
ABSTRACT
In the presence of a catalytic amount of (salen)Al(III)Cl complex (4e), reaction between alpha-isocyanoacetamides (1) and aldehydes (2) afforded the corresponding 5-aminooxazoles (3) in good yields and enantioselectivity.
Sigma-Aldrich
Cyanoacetamide, 99%
Sigma-Aldrich
(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride