MilliporeSigma
  • Highly beta-selective O-glucosidation due to the restricted twist-boat conformation.
Highly beta-selective O-glucosidation due to the restricted twist-boat conformation.
Organic letters (2007-03-16)
Yasunori Okada, Tatsuya Mukae, Kotaro Okajima, Miyoko Taira, Mari Fujita, Hidetoshi Yamada
ABSTRACT
[reaction: see text] Ethyl 1-thio-2,3,4,6-tetrakis-O-triisopropylsilyl-beta-d-glucopyranoside, ethyl 6-O-benzyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside, and ethyl 6-O-pivaloyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside induced highly beta-selective O-glucosidations. Among them, the 6-O-pivaloylated substrate provided the best selectivity up to alpha/beta = 3:97 with cyclohexylmethanol, and the substrate was used for glucosidations with secondary and tertiary alcohols in a highly beta-selective manner. The selectivity would be caused by the twist-boat conformation of the pyranose; this is the first beta-selective O-glucosidation based on conformational control of the pyranose ring.
Sigma-Aldrich
Cyclohexanemethanol, ReagentPlus®, ≥99%