MilliporeSigma
  • High-resolution nuclear magnetic resonance spectroscopy studies of polysaccharides crosslinked by sodium trimetaphosphate: a proposal for the reaction mechanism.
High-resolution nuclear magnetic resonance spectroscopy studies of polysaccharides crosslinked by sodium trimetaphosphate: a proposal for the reaction mechanism.
Carbohydrate research (2007-02-17)
Stéphane Lack, Virginie Dulong, Luc Picton, Didier Le Cerf, Eric Condamine
ABSTRACT
An NMR spectroscopy study ((31)P, (1)H, (13)C) of the postulated crosslinking mechanism of sodium trimetaphosphate (STMP) on polysaccharides is reported using methyl alpha-D-glucopyranoside as a model. In a first step, reaction of STMP with Glc-OMe gives grafted sodium tripolyphosphate (STPP(g)). On the one hand, STTP(g) can react with a second alcohol functionality to give a crosslinked monophosphate. On the other hand, a monophosphate (grafted phosphate) could be obtained by alkaline degradation of STPP(g). NMR spectroscopy allows to detect the various species formed and to obtain the crosslinking density of STMP-polysaccharides hydrogels.
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%