MilliporeSigma
  • Studies on three structurally related phenylenediamines with the mouse micronucleus assay system.
Studies on three structurally related phenylenediamines with the mouse micronucleus assay system.
Mutation research (1991-01-01)
L Soler-Niedziela, X Shi, J Nath, T Ong
ABSTRACT
Three structurally related compounds, 4-chloro-o-phenylenediamine (COP), 4-nitro-o-phenylenediamine (NOP) and p-phenylenediamine dihydrochloride (PPD), are used in fur dyes, inks and hair coloring formulations. COP has been reported to be carcinogenic in both rats and mice. NOP and PPD are non-carcinogens, but have consistently tested positive in short-term in vitro genotoxicity assays. Studies were undertaken to evaluate their genotoxicity with the in vivo mouse bone-marrow micronucleus assay. Five CD-1 male mice per dose were injected i.p. with the compounds and sacrificed at intervals of 24, 48 and 72 h. 2000 cells were scored per animal to determine the frequency of micronucleated-polychromatic erythrocytes (MPCE). COP induced significant dose-related increases in MPCE over the 3 doses tested at each of the sampling intervals. The peak response occurred at 24 h. No response was observed in animals treated with PPD or NOP.
Sigma-Aldrich
4-Chloro-o-phenylenediamine, 97%