MilliporeSigma
  • Herbicide effects on cuticle ultrastructure in Eleusine indica and Portulaca oleracea.
Herbicide effects on cuticle ultrastructure in Eleusine indica and Portulaca oleracea.
Biocell : official journal of the Sociedades Latinoamericanas de Microscopia Electronica ... et. al (2006-07-19)
Rosana N Malpassi
ABSTRACT
Eleusine indica and Portulaca oleracea are two common weeds in peanut crops in southern Córdoba. Two chemicals are frequently used to control them, quizalofop for grasses and lactofen for dicots. The objective is to study the effects of quizalofop and lactofen on cuticle ultrastructure in E. indica and P. oleracea, respectively. In the lab, quizalofop was applied on E. indica and lactofen on P. oleracea. Three plant categories were analyzed in each species: 3, 1-2, and no tiller in E. indica, and 8, 6, and 2 nomophylls in P. oleracea. Leaf samples from both species were collected at 7 and 16 days post-application and were treated for scanning electron microscopy. E. indica cuticle treated with lethal dose shows areas where epicuticular waxes disappear, specially in the youngest individuals. These areas are located predominantly on periclinal walls of typical epidermic cells and subsidiary cells. On the other hand, P. oleracea shows cuticle discontinuities that may be caused by lactofen entry. They are smaller and less frequent in plants having 8 or more nomophylls. The remaining waxes act as a herbicide accumulation compartment and, therefore, would partially prevent the active ingredient entry to epidermic cells.
Product Description
Supelco
Lactofen, PESTANAL®, analytical standard