MilliporeSigma
  • Mild and efficient oxidation of Hantzsch 1,4-dihydropyridines with sodium periodate catalyzed by a new polystyrene-bound Mn(TPP)Cl.
Mild and efficient oxidation of Hantzsch 1,4-dihydropyridines with sodium periodate catalyzed by a new polystyrene-bound Mn(TPP)Cl.
Bioorganic & medicinal chemistry letters (2006-01-24)
Majid Moghadam, Masoud Nasr-Esfahani, Shahram Tangestaninejad, Valiollah Mirkhani
ABSTRACT
Mild and efficient oxidation of Hantzsch 1,4-dihydropyridines with sodium periodate catalyzed by Mn(TTP)Cl supported on polystyrene-bound imidazole is reported. This heterogeneous catalyst is of great stability and reusability in the oxidation of 1,4-dihydropyridines with sodium periodate without significant loss of its catalytic activity.