MilliporeSigma
  • Efficient enzymatic synthesis of 4-methylumbelliferyl N-acetyllactosaminide and 4-methylumbelliferyl sialyl N-acetyllactosaminides employing beta-D-galactosidase and sialyltransferases.
Efficient enzymatic synthesis of 4-methylumbelliferyl N-acetyllactosaminide and 4-methylumbelliferyl sialyl N-acetyllactosaminides employing beta-D-galactosidase and sialyltransferases.
Biotechnology letters (2005-10-19)
Xiaoxiong Zeng, Yi Sun, Hirotaka Uzawa
ABSTRACT
4-Methylumbelliferyl N-acetyllactosaminide and 4-methylumbelliferyl sialyl N-acetyllactosaminides, which are used for the assay of sialytransferase, neuraminidase and fucosyltransferase, were synthesized, respectively, by the beta-D: -galactosidase from Bacillus circulans and by a recombinant rat alpha2,3-(N)-sialyltransferase or rat liver alpha2,6-(N)-sialyltransferase with CMP-N-acetylneuraminic acid as donor.
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, suitable for fluorescence, ≥99.0% (TLC)