MilliporeSigma
  • Hydrophobic drug delivery by self-assembling triblock copolymer-derived nanospheres.
Hydrophobic drug delivery by self-assembling triblock copolymer-derived nanospheres.
Biomacromolecules (2005-09-13)
Larisa Sheihet, Robert A Dubin, David Devore, Joachim Kohn
ABSTRACT
We describe the synthesis and characterization of a family of biocompatible ABA-triblock copolymers that comprised of hydrophilic A-blocks of poly(ethylene glycol) and hydrophobic B-blocks of oligomers of suberic acid and desaminotyrosyl-tyrosine esters. The triblock copolymers spontaneously self-assemble in aqueous solution into nanospheres, with hydrodynamic diameters between 40 and 70 nm, that do not dissociate under chromatographic and ultracentrifugation conditions. These nanospheres form strong complexes with hydrophobic molecules, including the fluorescent dye 5-dodecanoylaminofluorescein (DAF) and the antitumor drug, paclitaxel, but not with hydrophilic molecules such as fluorescein and Oregon Green. The nanosphere-paclitaxel complexes retain in vitro the high antiproliferative activity of paclitaxel, demonstrating that these nanospheres may be useful for delivery of the hydrophobic drugs.
Sigma-Aldrich
Suberic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Suberic acid, 98%