MilliporeSigma
  • Cinnamaldehyde and 2-methoxycinnamaldehyde as NF-kappaB inhibitors from Cinnamomum cassia.
Cinnamaldehyde and 2-methoxycinnamaldehyde as NF-kappaB inhibitors from Cinnamomum cassia.
Planta medica (2004-10-27)
Alavala Matta Reddy, Jee Hee Seo, Shi Yong Ryu, Yeong Shik Kim, Young Sup Kim, Kyung Rak Min, Youngsoo Kim
ABSTRACT
Nuclear factor (NF)-cB is a transcription factor regulating the expression of inflammatory and immune genes. In the present study, an extract from stem bark of Cinnamomum cassia Blume(Lauraceae) was discovered to have an inhibitory effect on LPS-induced NF-KB transcriptional activity, which was determined using macrophages RAW 264.7 transfected stably with an alkaline phosphatase reporter construct containing four copies of the NF-KB binding KB sequence. Following activity-guided fractionation, trans-cinnamaldehyde and 2-methoxycinnamaldehyde were identified as the NF-KB inhibitors from C cassia with IC50 values of 43 MM and 31 pM, respectively. As a positive control, caffeic acid phenethyl ester (CAPE) showed an IC50 value of 2 uM on NF-KB transcriptional activity. Both trans-cinnamaldehyde and 2-methoxycinnamaldehyde inhibited LPS-induced DNA binding activity of NF-KB in addition to NF-KB transcription-al activity.
Sigma-Aldrich
2-Methoxycinnamaldehyde, natural, 98%, FG