MilliporeSigma
A benzenoid from the stem of Acanthopanax senticosus.
Archives of pharmacal research (2004-10-12)
Jiyoung Ryu, Dongwook Son, Jungil Kang, Hyun-Su Kim, Bak-Kwang Kim, Sanghyun Lee
ABSTRACT
Seven compounds were isolated from the stem of Acanthopanax senticosus by repeated column chromatography. Their structures were elucidated as isovanillin (1), (-)-sesamin (2), isofraxidin (3), (+)-syringaresinol (4), 5-hydroxymethylfurfural (5), eleutheroside B (6), and eleutheroside E (7) by spectral analysis. Among them, isovanillin (1) was isolated for the first time from the family Araliaceae.
Sigma-Aldrich
3-Hydroxy-4-methoxybenzaldehyde, 99%
Sigma-Aldrich
Isovanillin, ≥95.0%