MilliporeSigma
  • Mineral induced phosphorylation of glycolate ion--a metaphor in chemical evolution.
Mineral induced phosphorylation of glycolate ion--a metaphor in chemical evolution.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2001-09-07)
V Kolb, S Zhang, Y Xu, G Arrhenius
ABSTRACT
Bilateral surface-active minerals with excess positive charge concentrate glycolate and trimetaphosphate ion from l0(-3) m aqueous solution to half-saturation of the internal surface sites, and induce phosphorylation of glycolate ion in the mineral with trimetaphosphate, sorbed from l0(-2) m solution. By utilizing reactants from dilute solution at near-neutral pH, and eliminating the need for participating organic nitrogen compounds, the reaction comprises several elements considered necessary for geochemical realism in models for molecular evolution.
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%