MilliporeSigma
  • Small molecule inhibitor of mitotic spindle bipolarity identified in a phenotype-based screen.
Small molecule inhibitor of mitotic spindle bipolarity identified in a phenotype-based screen.
Science (New York, N.Y.) (1999-11-05)
T U Mayer, T M Kapoor, S J Haggarty, R W King, S L Schreiber, T J Mitchison
ABSTRACT
Small molecules that perturb specific protein functions are valuable tools for dissecting complex processes in mammalian cells. A combination of two phenotype-based screens, one based on a specific posttranslational modification, the other visualizing microtubules and chromatin, was used to identify compounds that affect mitosis. One compound, here named monastrol, arrested mammalian cells in mitosis with monopolar spindles. In vitro, monastrol specifically inhibited the motility of the mitotic kinesin Eg5, a motor protein required for spindle bipolarity. All previously known small molecules that specifically affect the mitotic machinery target tubulin. Monastrol will therefore be a particularly useful tool for studying mitotic mechanisms.
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid