MilliporeSigma
  • A soluble activin type IIA receptor induces bone formation and improves skeletal integrity.
A soluble activin type IIA receptor induces bone formation and improves skeletal integrity.
Proceedings of the National Academy of Sciences of the United States of America (2008-05-08)
R Scott Pearsall, Ernesto Canalis, Milton Cornwall-Brady, Kathryn W Underwood, Brendan Haigis, Jeffrey Ucran, Ravindra Kumar, Eileen Pobre, Asya Grinberg, Eric D Werner, Vaida Glatt, Lisa Stadmeyer, Deanna Smith, Jasbir Seehra, Mary L Bouxsein
ABSTRACT
Diseases that affect the regulation of bone turnover can lead to skeletal fragility and increased fracture risk. Members of the TGF-beta superfamily have been shown to be involved in the regulation of bone mass. Activin A, a TGF-beta signaling ligand, is present at high levels in bone and may play a role in the regulation of bone metabolism. Here we demonstrate that pharmacological blockade of ligand signaling through the high affinity receptor for activin, type II activin receptor (ActRIIA), by administration of the soluble extracellular domain of ActRIIA fused to a murine IgG2a-Fc, increases bone formation, bone mass, and bone strength in normal mice and in ovariectomized mice with established bone loss. These observations support the development of this pharmacological strategy for the treatment of diseases with skeletal fragility.
Sigma-Aldrich
Formalin Free Tissue Fixative
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture