MilliporeSigma
  • Gel-eluted liquid fraction entrapment electrophoresis: an electrophoretic method for broad molecular weight range proteome separation.
Gel-eluted liquid fraction entrapment electrophoresis: an electrophoretic method for broad molecular weight range proteome separation.
Analytical chemistry (2008-01-31)
John C Tran, Alan A Doucette
ABSTRACT
Although well-established as a technique for protein purification, the application of continuous elution tube gel electrophoresis to proteome fractionation remains problematic. Difficulties associated with sample collection, particularly at the high mass range or at low sample loadings, continue to plague the technique. Furthermore, an upper mass limit is imposed as slow-moving higher molecular weight proteins are progressively diluted during the collection phase. In short, with current technology, effective separation over a broad mass range has not been achieved. In this work, we present improved techniques for continuous elution tube gel electrophoresis to accommodate broad mass range separation of proteins. Our device enables rapid partitioning of a proteome into discrete mass range fractions in the solution phase. High recovery is achieved at submicrogram to milligram sample loadings. We demonstrate comprehensive, reproducible separations of protein mixtures, as well as separation of a proteome in as fast as 1 h, over mass ranges from below 10 to 250 kDa. Finally, we identified proteins from a prefractionated standard protein mixture using liquid chromatography tandem mass spectrometric (LC-MS/MS) analysis.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein