MilliporeSigma
  • GATA2-induced silencing and LIM-homeodomain protein-induced activation are mediated by a bi-functional response element in the rat GnRH receptor gene.
GATA2-induced silencing and LIM-homeodomain protein-induced activation are mediated by a bi-functional response element in the rat GnRH receptor gene.
Molecular endocrinology (Baltimore, Md.) (2012-12-06)
Anne-Laure Schang, Anne Granger, Bruno Quérat, Christian Bleux, Joëlle Cohen-Tannoudji, Jean-Noël Laverrière
ABSTRACT
GATA2 transcription factor and LIM homeodomain proteins Islet1 (ISL1) and LIM homeobox 3 (LHX3) are suspected to be involved in gonadotrope cell fate and maintenance. The GnRH receptor gene (Gnrhr), crucial for gonadotrope function, is expressed in the pituitary gland from embryonic day 13.5 onward, well before LH and FSH β-subunits. This expression pattern together with the presence of WGATAR and TAAT motifs in Gnrhr promoter sequences suggests the involvement of early transcription factors in promoter activation. In this study, using a well-characterized transgenic mouse model, GATA2 was found colocalized with Gnrhr promoter activity in the pituitary. Transient transfection of Gnrhr promoter luciferase fusion constructs together with either GATA2 expression vectors or small interfering RNA in gonadotrope cell lines indicated that GATA2, which typically acts as a trans-activator, unexpectedly repressed Gnrhr promoter activity. Using DNA chromatography affinity and EMSA, we demonstrated that GATA2 operates via a response element containing a peculiar palindromic GATA motif that overlaps a critical TAAT motif involved in LHX3/ISL1 trans-activation. Indeed, despite the inhibitory action of GATA2, this element displayed a clear-cut enhancer activity in gonadotrope cells. Chromatin immunoprecipitation assays indicated that GATA2, LHX3, and ISL1 interact with a Gnrhr promoter fragment encompassing this element. The trans-repressive action of GATA2 on Gnrhr promoter activity is likely balanced or even hindered by trans-activating effects of LIM homeodomain proteins via this novel bifunctional LIM/GATA response element. Such a hierarchical interplay may contribute to finely adjust Gnrhr gene expression in gonadotrope cell lineage during pituitary development as well as in the adult animal.
Sigma-Aldrich
Anti-LIM-3 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MMP-1 Antibody, hinge region, from rabbit, purified by affinity chromatography