MilliporeSigma
  • Nitrate respiratory metabolism in an obligately autotrophic hydrogen-oxidizing bacterium, Hydrogenobacter thermophilus TK-6.
Nitrate respiratory metabolism in an obligately autotrophic hydrogen-oxidizing bacterium, Hydrogenobacter thermophilus TK-6.
Archives of microbiology (2001-03-29)
M Suzuki, Z J Cui, M Ishii, Y Igarashi
ABSTRACT
Hydrogenobacter thermophilus strain TK-6 was observed to grow anaerobically on nitrate as an electron acceptor when molecular hydrogen was used as an energy source. Nitrite was detected as the product of a respiratory reaction. 15NO, 15N2O, and 15N2 were detected with Na15NO3 as an electron acceptor. Western immunoblot analysis showed that cell-free extracts from cells grown on nitrate reacted with antibodies against heme cd1-type nitrite reductase from Pseudomonas aeruginosa. The positive bands, which had molecular masses similar to that of the heme cd1-type nitrite reductase, were also stained by heme staining. These results indicate that nitrite reductase of strain TK-6 is a heme cd1-type enzyme. Activity of ATP:citrate lyase, one of the key enzymes of the reductive TCA cycle, was detected in cell-free extract of cells cultivated on nitrate, which indicates that the cycle operates during anaerobic growth.
Sigma-Aldrich
Citrate Lyase from Klebsiella pneumoniae, lyophilized powder, ≥0.20 unit/mg solid