MilliporeSigma
  • A pathologically expanded, clonal lineage of IL-21-producing CD4+ T cells drives inflammatory neuropathy.
A pathologically expanded, clonal lineage of IL-21-producing CD4+ T cells drives inflammatory neuropathy.
The Journal of clinical investigation (2024-08-01)
Maryamsadat Seyedsadr, Madison F Bang, Ethan C McCarthy, Shirley Zhang, Ho-Chung Chen, Mahnia Mohebbi, Willy Hugo, Jason K Whitmire, Melissa G Lechner, Maureen A Su
ABSTRACT
Inflammatory neuropathies, which include chronic inflammatory demyelinating polyneuropathy (CIDP) and Guillain Barré syndrome (GBS), result from autoimmune destruction of the PNS and are characterized by progressive weakness and sensory loss. CD4+ T cells play a key role in the autoimmune destruction of the PNS. Yet, key properties of pathogenic CD4+ T cells remain incompletely understood. Here, we used paired single-cell RNA-Seq (scRNA-Seq) and single-cell T cell receptor-sequencing (scTCR-Seq) of peripheral nerves from an inflammatory neuropathy mouse model to identify IL-21-expressing CD4+ T cells that were clonally expanded and multifunctional. These IL-21-expressing CD4+ T cells consisted of 2 transcriptionally distinct expanded cell populations, which expressed genes associated with T follicular helper (Tfh) and T peripheral helper (Tph) cell subsets. Remarkably, TCR clonotypes were shared between these 2 IL-21-expressing cell populations, suggesting a common lineage differentiation pathway. Finally, we demonstrated that IL-21 receptor-KO (IL-21R-KO) mice were protected from neuropathy development and had decreased immune infiltration into peripheral nerves. IL-21 signaling upregulated CXCR6, a chemokine receptor that promotes CD4+ T cell localization in peripheral nerves. Together, these findings point to IL-21 signaling, Tfh/Tph differentiation, and CXCR6-mediated cellular localization as potential therapeutic targets in inflammatory neuropathies.
Sigma-Aldrich
BCL6 Inhibitor, 79-6, The BCL6 Inhibitor, 79-6 controls the biological activity of BCL6. This small molecule/inhibitor is primarily used for Cancer applications.