MilliporeSigma
  • Perfluorooctane sulfonic acid (PFOS) inhibits vessel formation in a human 3D co-culture angiogenesis model (NCFs/HUVECs).
Perfluorooctane sulfonic acid (PFOS) inhibits vessel formation in a human 3D co-culture angiogenesis model (NCFs/HUVECs).
Environmental pollution (Barking, Essex : 1987) (2021-11-21)
Martin Forsthuber, Raimund Widhalm, Sebastian Granitzer, Andreas Marius Kaiser, Hanns Moshammer, Markus Hengstschläger, Helmut Dolznig, Claudia Gundacker
ABSTRACT
Perfluorooctane sulfonic acid (PFOS) is a ubiquitous environmental pollutant. In humans, PFOS exposure has been associated with a number of adverse health outcomes, including reduced birth weight. Whether PFOS is capable of affecting angiogenesis and thus possibly fetal development is unknown. Therefore, we investigated 1) the metabolic activity of PFOS-exposed endothelial cells (human umbilical vein endothelial cells, HUVECs), fibroblasts (normal colon fibroblasts, NCFs), and epithelial cells (human colorectal carcinoma cells, HCT116), 2) PFOS-specific inhibition of vascular endothelial growth factor receptor (VEGFR)2 stimulation in KDR/NFAT-RE HEK293 cells, and 3) the antiangiogenic potential of PFOS in a 3D in vitro angiogenesis model of HUVECs and NCFs. In terms of metabolic activity, endothelial cells (HUVECs) were much more sensitive to PFOS than fibroblasts (NCFs) or epithelial cells (HCT116). VEGFR2 signaling in KDR/NFAT-RE HEK293 cells decreased with increasing PFOS concentrations. In co-culture (angiogenesis assay), PFOS treatment resulted in a dose-dependent reduction in tip and branch formation, tip length (μm), and total structural area (μm2) with stable metabolic activity of HUVECs up to high concentrations. We conclude that PFOS possesses antiangiogenic properties. Inhibition of VEGFR2 signaling indicates a possible mechanism of action that can be linked to an existing Adverse Outcome Pathway (AOP43) containing the AO reduced birth weight. Further studies are needed to confirm PFOS-specific adverse effects on angiogenesis, placental perfusion, and fetal growth.
Sigma-Aldrich
Staurosporine from Streptomyces sp., for molecular biology, ≥95% (HPLC)