MilliporeSigma
Software-assisted serum metabolite quantification using NMR.
Analytica chimica acta (2016-08-11)
Young-Sang Jung, Jin-Seong Hyeon, Geum-Sook Hwang
ABSTRACT
The goal of metabolomics is to analyze a whole metabolome under a given set of conditions, and accurate and reliable quantitation of metabolites is crucial. Absolute concentration is more valuable than relative concentration; however, the most commonly used method in NMR-based serum metabolic profiling, bin-based and full data point peak quantification, provides relative concentration levels of metabolites and are not reliable when metabolite peaks overlap in a spectrum. In this study, we present the software-assisted serum metabolite quantification (SASMeQ) method, which allows us to identify and quantify metabolites in NMR spectra using Chenomx software. This software uses the ERETIC2 utility from TopSpin to add a digitally synthesized peak to a spectrum. The SASMeQ method will advance NMR-based serum metabolic profiling by providing an accurate and reliable method for absolute quantification that is superior to bin-based quantification.
Product Description
Bruker® SampleJet NMR tubes with caps, L 103.5 mm, O.D. 5.0 mm, cap