MilliporeSigma
  • Limiting Monoamines Degradation Increases L-DOPA Pro-Locomotor Action in Newborn Rats.
Limiting Monoamines Degradation Increases L-DOPA Pro-Locomotor Action in Newborn Rats.
International journal of molecular sciences (2023-10-14)
Inès Khsime, Marie Boulain, Abderrahman Fettah, Abdeslam Chagraoui, Gilles Courtand, Philippe De Deurwaerdère, Laurent Juvin, Grégory Barrière
ABSTRACT
L-DOPA, the precursor of catecholamines, exerts a pro-locomotor action in several vertebrate species, including newborn rats. Here, we tested the hypothesis that decreasing the degradation of monoamines can promote the pro-locomotor action of a low, subthreshold dose of L-DOPA in five-day-old rats. The activity of the degrading pathways involving monoamine oxidases or catechol-O-methyltransferase was impaired by injecting nialamide or tolcapone, respectively. At this early post-natal stage, the capacity of the drugs to trigger locomotion was investigated by monitoring the air-stepping activity expressed by the animals suspended in a harness above the ground. We show that nialamide (100 mg/kg) or tolcapone (100 mg/kg), without effect on their own promotes maximal expression of air-stepping sequences in the presence of a sub-effective dose of L-DOPA (25 mg/kg). Tissue measurements of monoamines (dopamine, noradrenaline, serotonin and some of their metabolites) in the cervical and lumbar spinal cord confirmed the regional efficacy of each inhibitor toward their respective enzyme. Our experiments support the idea that the raise of monoamines boost L-DOPA's locomotor action. Considering that both inhibitors differently altered the spinal monoamines levels in response to L-DOPA, our data also suggest that maximal locomotor response can be reached with different monoamines environment.
Sigma-Aldrich
Tolcapone, ≥98% (HPLC)