MilliporeSigma
  • Occludin is overexpressed in tubo-ovarian high-grade serous carcinoma compared to mesothelioma and is a marker of tumor progression and chemoresistance.
Occludin is overexpressed in tubo-ovarian high-grade serous carcinoma compared to mesothelioma and is a marker of tumor progression and chemoresistance.
Clinical & experimental metastasis (2023-12-23)
Margarida Varela Dos Santos, Arild Holth, Katharina Bischof, Ben Davidson
ABSTRACT
The objective of this study was to analyze the expression and prognostic role of the tight junction protein occludin in high-grade serous carcinoma (HGSC). Occludin protein expression by immunohistochemistry was analyzed in 602 HGSC (417 effusions, 185 surgical specimens). Expression in mesothelioma (n = 87; 45 effusions, 42 surgical specimens) was studied for comparative purposes. Occludin protein expression was found in 587/602 (98%) HGSC vs. 40/87 (46%) mesotheliomas and was predominantly limited to < 5% of cells in the latter (p < 0.001). Occludin was additionally overexpressed in HGSC effusions compared to surgical specimens (p < 0.001) and was overexpressed in post-chemotherapy effusions compared to chemo-naive effusions tapped at diagnosis (p = 0.015). Occludin expression in HGSC surgical specimens was associated with poor chemoresponse (p < 0.001) and primary resistance (p = 0.001). Expression in effusions and surgical specimens was unrelated to survival (p > 0.05). In conclusion, occludin expression is higher in HGSC compared to mesothelioma, and this protein is overexpressed in HGSC effusions, possibly reflecting changes in adhesion related to anchorage-independent growth in this microenvironment. Overexpression in post-chemotherapy compared to chemo-naïve effusions suggest a role in disease progression. Occludin expression in surgical specimens may be related to chemoresistance.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 31C, purified immunoglobulin, buffered aqueous solution