MilliporeSigma
  • Seroepidemiology and risk factors for SARS-CoV-2 infection among household members of food processing and farm workers in North Carolina.
Seroepidemiology and risk factors for SARS-CoV-2 infection among household members of food processing and farm workers in North Carolina.
IJID regions (2023-04-11)
Michael Sciaudone, Melissa K Cutshaw, Colleen M McClean, Roberto Lacayo, Oksana Kharabora, Katherine Murray, Stephen Strohminger, Miriana Moreno Zivanovich, Rachel Gurnett, Alena J Markmann, Emperatriz Morales Salgado, D Ryan Bhowmik, Edwin Castro-Arroyo, Ross M Boyce, Allison E Aiello, David Richardson, Jonathan J Juliano, Natalie M Bowman
ABSTRACT
Racial and ethnic minorities have borne a disproportionate burden from coronavirus disease 2019 (COVID-19). Certain essential occupations, including food processing and farm work, employ large numbers of Hispanic migrant workers and have been shown to carry an especially high risk of infection. This observational cohort study measured the seroprevalence of severe acute respiratory syndrome coronavirus-2 (SARS-CoV-2) and assessed the risk factors for seropositivity among food processing and farm workers, and members of their households, in North Carolina, USA. Participants completed questionnaires, blood samples were collected, and an enzyme-linked immunosorbent assay was used to assess SARS-CoV-2 seropositivity. Univariate and multi-variate analyses were undertaken to identify risk factors associated with seropositivity, using generalized estimating equations to account for household clustering. Among the 218 participants, 94.5% were Hispanic, and SARS-CoV-2 seropositivity was 50.0%. Most seropositive individuals did not report a history of illness compatible with COVID-19. Attending church, having a prior history of COVID-19, having a seropositive household member, and speaking Spanish as one's primary language were associated with SARS-CoV-2 seropositivity, while preventive behaviours were not. These findings underscore the substantial burden of COVID-19 among a population of mostly Hispanic essential workers and their households in rural North Carolina. This study contributes to a large body of evidence showing that Hispanic Americans have suffered a disproportionate burden of COVID-19. This study also highlights the epidemiologic importance of viral transmission within the household.
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution