MilliporeSigma
  • High throughput compound screening in neuronal cells identifies statins as activators of ataxin 3 expression.
High throughput compound screening in neuronal cells identifies statins as activators of ataxin 3 expression.
Scientific reports (2023-09-10)
Fabian Stahl, Ina Schmitt, Philip Denner, Laura de Boni, Ullrich Wüllner, Peter Breuer
ABSTRACT
The spinocerebellar ataxias (SCA) comprise a group of inherited neurodegenerative diseases. SCA3 is the most common form, caused by the expansion of CAG repeats within the ataxin 3 (ATXN3) gene. The mutation results in the expression of an abnormal protein, containing long polyglutamine (polyQ) stretches. The polyQ stretch confers a toxic gain of function and leads to misfolding and aggregation of ATXN3 in neurons. Thus, modulators of ATXN3 expression could potentially ameliorate the pathology in SCA3 patients. Therefore, we generated a CRISPR/Cas9 modified ATXN3-Exon4-Luciferase (ATXN3-LUC) genomic fusion- and control cell lines to perform a reporter cell line-based high-throughput screen comprising 2640 bioactive compounds, including the FDA approved drugs. We found no unequivocal inhibitors of, but identified statins as activators of the LUC signal in the ATXN3-LUC screening cell line. We further confirmed that Simvastatin treatment of wild type SK-N-SH cells increases ATXN3 mRNA and protein levels which likely results from direct binding of the activated sterol regulatory element binding protein 1 (SREBP1) to the ATXN3 promotor. Finally, we observed an increase of normal and expanded ATXN3 protein levels in a patient-derived cell line upon Simvastatin treatment, underscoring the potential medical relevance of our findings.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-HMG-CoA reductase Antibody, from rabbit, purified by affinity chromatography