MilliporeSigma
  • Whole-genome screens reveal regulators of differentiation state and context-dependent migration in human neutrophils.
Whole-genome screens reveal regulators of differentiation state and context-dependent migration in human neutrophils.
Nature communications (2023-09-19)
Nathan M Belliveau, Matthew J Footer, Emel Akdoǧan, Aaron P van Loon, Sean R Collins, Julie A Theriot
ABSTRACT
Neutrophils are the most abundant leukocyte in humans and provide a critical early line of defense as part of our innate immune system. We perform a comprehensive, genome-wide assessment of the molecular factors critical to proliferation, differentiation, and cell migration in a neutrophil-like cell line. Through the development of multiple migration screen strategies, we specifically probe directed (chemotaxis), undirected (chemokinesis), and 3D amoeboid cell migration in these fast-moving cells. We identify a role for mTORC1 signaling in cell differentiation, which influences neutrophil abundance, survival, and migratory behavior. Across our individual migration screens, we identify genes involved in adhesion-dependent and adhesion-independent cell migration, protein trafficking, and regulation of the actomyosin cytoskeleton. This genome-wide screening strategy, therefore, provides an invaluable approach to the study of neutrophils and provides a resource that will inform future studies of cell migration in these and other rapidly migrating cells.
Sigma-Aldrich
Anti-phospho-p70 S6 Kinase (Thr389) Antibody, from rabbit, purified by affinity chromatography