MilliporeSigma
  • STING controls T cell memory fitness during infection through T cell-intrinsic and IDO-dependent mechanisms.
STING controls T cell memory fitness during infection through T cell-intrinsic and IDO-dependent mechanisms.
Proceedings of the National Academy of Sciences of the United States of America (2023-01-13)
Michael J Quaney, Curtis J Pritzl, Dezzarae Luera, Rebecca J Newth, Karin M Knudson, Vikas Saxena, Caitlyn Guldenpfennig, Diana Gil, Chris S Rae, Peter Lauer, Mark A Daniels, Emma Teixeiro
ABSTRACT
Stimulator of interferon genes (STING) signaling has been extensively studied in inflammatory diseases and cancer, while its role in T cell responses to infection is unclear. Using Listeria monocytogenes strains engineered to induce different levels of c-di-AMP, we found that high STING signals impaired T cell memory upon infection via increased Bim levels and apoptosis. Unexpectedly, reduction of TCR signal strength or T cell-STING expression decreased Bim expression, T cell apoptosis, and recovered T cell memory. We found that TCR signal intensity coupled STING signal strength to the unfolded protein response (UPR) and T cell survival. Under strong STING signaling, Indoleamine-pyrrole 2,3-dioxygenase (IDO) inhibition also reduced apoptosis and led to a recovery of T cell memory in STING sufficient CD8 T cells. Thus, STING signaling regulates CD8 T cell memory fitness through both cell-intrinsic and extrinsic mechanisms. These studies provide insight into how IDO and STING therapies could improve long-term T cell protective immunity.
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%