MilliporeSigma
  • lncRNA UCA1 regulates miR-132/Lrrfip1 axis to promote vascular smooth muscle cell proliferation.
lncRNA UCA1 regulates miR-132/Lrrfip1 axis to promote vascular smooth muscle cell proliferation.
Open medicine (Warsaw, Poland) (2023-08-03)
Wenming Chen, Wei Zhao, Minghui Hao, Yuping Wang
ABSTRACT
UCA1 is predicted to bind to miR-132, which is a key player in the proliferation of vascular smooth muscle cells (VSMCs). This research studied the role of lncRNA UCA1 in atherosclerosis. The binding of UCA1 to miR-132 was proved by dual luciferase activity assay and RNA immunoprecipitation. UCA1 and miR-132 failed to affect each other's expression in VSMCs. UCA1 was upregulated and miR-132 was decreased in atherosclerosis plasma. However, they are not closely correlated across atherosclerosis and control plasma sample. Interestingly, UCA1 suppressed the role of miR-132 in downregulating Lrrfip1 expression and promoting VSMC proliferation. Therefore, UCA1 is downregulated in atherosclerosis and may regulate miR-132/Lrrfip1 axis to promote VSMC proliferation.
Sigma-Aldrich
Human Aortic Smooth Muscle Cells: HAOSMC, adult