MilliporeSigma
  • An Artemisia scoparia extract attenuates glucocorticoid-induced lipolysis in adipocytes.
An Artemisia scoparia extract attenuates glucocorticoid-induced lipolysis in adipocytes.
Obesity (Silver Spring, Md.) (2023-05-31)
Innocence Harvey, Jacqueline M Stephens
ABSTRACT
Prescription glucocorticoid (GC) use is widespread across developed countries for the treatment of several inflammatory conditions. Elevated GCs are known to promote lipolysis and metabolic disorders. An extract of Artemisia scoparia (SCO) has been shown to reduce lipolysis and promote metabolic health but has not been investigated in the context of excess GCs. Our aim was to examine the effects of SCO on GC-induced lipolysis. Mature adipocytes were pretreated with vehicle or SCO, then exposed to either the synthetic GC dexamethasone (DEX) or tumor necrosis factor alpha (TNFα). Medium was collected and assayed for glycerol and fatty acids as measures of lipolysis. The expression of several lipolytic genes and proteins was assessed, and the involvement of glucocorticoid receptor (GR) in SCO's effects was also interrogated. SCO significantly attenuated DEX-induced lipolysis but did not interfere with DEX-mediated changes in inflammatory gene profiles in adipocytes. SCO treatment resulted in significant reductions in monomeric phosphodiesterase (PDE) protein levels while elevating PDE multimeric complex formation, but other canonical lipolytic mediators were unaltered. SCO attenuated lipolysis even when GR expression was significantly knocked down. Finally, it was demonstrated that SCO was distinct from rosiglitazone in its antilipolytic effects. SCO attenuates GC-induced lipolysis independently of GR activity. Future studies are needed to elucidate underlying mechanisms.
Sigma-Aldrich
Roflumilast, ≥98% (HPLC)