MilliporeSigma
  • PP1γ regulates neuronal insulin signaling and aggravates insulin resistance leading to AD-like phenotypes.
PP1γ regulates neuronal insulin signaling and aggravates insulin resistance leading to AD-like phenotypes.
Cell communication and signaling : CCS (2023-04-22)
Yamini Yadav, Medha Sharma, Chinmoy Sankar Dey
ABSTRACT
PP1γ is one of the isoforms of catalytic subunit of a Ser/Thr phosphatase PP1. The role of PP1γ in cellular regulation is largely unknown. The present study investigated the role of PP1γ in regulating neuronal insulin signaling and insulin resistance in neuronal cells. PP1 was inhibited in mouse neuroblastoma cells (N2a) and human neuroblastoma cells (SH-SY5Y). The expression of PP1α and PP1γ was determined in insulin resistant N2a, SH-SY5Y cells and in high-fat-diet-fed-diabetic mice whole-brain-lysates. PP1α and PP1γ were silenced by siRNA in N2a and SH-SY5Y cells and effect was tested on AKT isoforms, AS160 and GSK3 isoforms using western immunoblot, GLUT4 translocation by confocal microscopy and glucose uptake by fluorescence-based assay. Results showed that, in one hand PP1γ, and not PP1α, regulates neuronal insulin signaling and insulin resistance by regulating phosphorylation of AKT2 via AKT2-AS160-GLUT4 axis. On the other hand, PP1γ regulates phosphorylation of GSK3β via AKT2 while phosphorylation of GSK3α via MLK3. Imbalance in this regulation results into AD-like phenotype. PP1γ acts as a linker, regulating two pathophysiological conditions, neuronal insulin resistance and AD. Video Abstract.
Sigma-Aldrich
FK-506, Streptomyces sp., Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2.