MilliporeSigma
  • A novel function for the sperm adhesion protein IZUMO1 in cell-cell fusion.
A novel function for the sperm adhesion protein IZUMO1 in cell-cell fusion.
The Journal of cell biology (2022-11-18)
Nicolas G Brukman, Kohdai P Nakajima, Clari Valansi, Kateryna Flyak, Xiaohui Li, Tetsuya Higashiyama, Benjamin Podbilewicz
ABSTRACT
Mammalian sperm-egg adhesion depends on the trans-interaction between the sperm-specific type I glycoprotein IZUMO1 and its oocyte-specific GPI-anchored receptor JUNO. However, the mechanisms and proteins (fusogens) that mediate the following step of gamete fusion remain unknown. Using live imaging and content mixing assays in a heterologous system and structure-guided mutagenesis, we unveil an unexpected function for IZUMO1 in cell-to-cell fusion. We show that IZUMO1 alone is sufficient to induce fusion, and that this ability is retained in a mutant unable to bind JUNO. On the other hand, a triple mutation in exposed aromatic residues prevents this fusogenic activity without impairing JUNO interaction. Our findings suggest a second function for IZUMO1 as a unilateral mouse gamete fusogen.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-IZUMO1 Antibody, clone Mab120, clone Mab120, from rat
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~5,000 IU