MilliporeSigma
  • Blockade of TMEM16A protects against renal fibrosis by reducing intracellular Cl- concentration.
Blockade of TMEM16A protects against renal fibrosis by reducing intracellular Cl- concentration.
British journal of pharmacology (2021-12-29)
Xiao-Long Li, Jing Liu, Xiao-Shan Chen, Li-Min Cheng, Wei-Ling Liu, Xing-Feng Chen, Yue-Jiao Li, Yong-Yuan Guan, Xin Zeng, Yan-Hua Du
ABSTRACT
Renal fibrosis is the final common outcome in most forms of chronic kidney disease (CKD). However, the underlying causal mechanisms remain obscure. The present study examined whether transmembrane member 16A (TMEM16A), a Ca2+ -activated chloride channel, contributes to the progression of renal fibrosis. Masson staining, western blot and immunohistochemistry were used to measure renal fibrosis and related proteins expression. MQAE was used to evaluate the intracellular Cl- concentration. TMEM16A expression was significantly up-regulated in fibrotic kidneys of unilateral ureteral obstruction (UUO) and high-fat diet murine models and in renal samples of IgA nephropathy patients. In vivo knockdown of TMEM16A with adenovirus harbouring TMEM16A-shRNA or inhibition of TMEM16A channel activity with inhibitors CaCCinh-A01 or T16Ainh-A01 effectively prevented UUO-induced renal fibrosis and decreased protein expression of fibronectin, α-SMA and collagen in the obstructed kidneys. In cultured HK2 cells, knockdown or inhibition of TMEM16A suppressed TGF-β1-induced epithelial-mesenchymal transition, reduced snail1 expression and phosphorylation of Smad2/3 and ERK1/2, whereas overexpression of TMEM16A showed the opposite effects. TGF-β1 increased [Cl- ]i in HK2 cells, which was inhibited by knockdown or inhibition of TMEM16A. Reducing [Cl- ]i significantly blunted TGF-β1-induced Smad2/3 phosphorylation and profibrotic factors expression. The profibrotic effects of TGF-β1 were also reduced by inhibition of serum- and glucocorticoid-inducible protein kinase 1 (SGK1). SGK1 was also suppressed by reducing [Cl- ]i. Blockade of TMEM16A prevented the progression of kidney fibrosis, likely by suppressing [Cl- ]i/SGK1/TGF-β1 signalling pathway. TMEM16A may be a potential new therapeutic target against renal fibrosis.
Sigma-Aldrich
Calcium-activated Chloride Channel Inhibitor, CaCCinh-A01, The Calcium-activated Chloride Channel Inhibitor, CaCCinh-A01 controls the biological activity of Calcium-activated Chloride Channel.