MilliporeSigma
  • Interfering with lipid metabolism through targeting CES1 sensitizes hepatocellular carcinoma for chemotherapy.
Interfering with lipid metabolism through targeting CES1 sensitizes hepatocellular carcinoma for chemotherapy.
JCI insight (2022-12-07)
Gang Li, Xin Li, Iqbal Mahmud, Jazmin Ysaguirre, Baharan Fekry, Shuyue Wang, Bo Wei, Kristin L Eckel-Mahan, Philip L Lorenzi, Richard Lehner, Kai Sun
ABSTRACT
Hepatocellular carcinoma (HCC) is the most common lethal form of liver cancer. Apart from surgical removal and transplantation, other treatments have not yet been well established for patients with HCC. Herein, we found that carboxylesterase 1 (CES1) was expressed at various levels in HCC. We further revealed that blockage of CES1 by pharmacological and genetical approaches leads to altered lipid profiles that are directly linked to impaired mitochondrial function. Mechanistically, LC-MS/MS and lipidomic analyses revealed that lipid signaling molecules, including polyunsaturated fatty acids (PUFAs), which activate PPARα/γ, were dramatically reduced upon CES1 inhibition. As a result, SCD, a PPARα/γ target gene involved in tumor progression and chemoresistance, was significantly downregulated. Consistently, clinical analysis demonstrated a strong correlation between the protein levels of CES1 and SCD in HCC. Interference with lipid signaling by targeting the "CES1-PPARα/γ-SCD" axis sensitized HCC cells to cisplatin treatment. As a demonstration, the growth of HCC xenograft tumors in NU/J mice was potently limited by co-administration of cisplatin and CES1 inhibition. Our results suggest that CES1 is a promising therapeutic target for HCC treatment.
Sigma-Aldrich
Anti-SCD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CES1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution