MilliporeSigma
Engineering Antibodies on the Surface of CHO Cells.
Methods in molecular biology (Clifton, N.J.) (2019-10-19)
Annalee W Nguyen, Kevin Le, Jennifer A Maynard
ABSTRACT
While antibody libraries are traditionally screened in phage, bacterial, or yeast display formats, they are produced in large scale for pharmaceutical and commercial use in mammalian cell lines. The simpler organisms used for screening have significantly different folding and glycosylation machinery than mammalian cells; consequently, clones resulting from these libraries may require further optimization for mammalian cell expression. To streamline the antibody discovery process, we developed a Chinese hamster ovary (CHO) cell-based selection system that allows for long-term display of antibody Fab fragments. This system is facilitated by a semi-stable Epi-CHO episomal platform to maintain antibody expression for up to 2 months and is compatible with standard PCR-based mutagenesis strategies. This protocol describes the simple and accessible use of CHO display coupled with flow cytometry to enrich for antibody variants with increased ligand-binding affinity from large libraries of ~106 variants, using HER2-binding antibodies as an example.
Sigma-Aldrich
Anti-Human Kappa Light Chain (Bound and Free)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution