MilliporeSigma
  • Proteolysis and changes in meat quality of chicken pectoralis major and iliotibialis muscles in relation to muscle fiber type distribution.
Proteolysis and changes in meat quality of chicken pectoralis major and iliotibialis muscles in relation to muscle fiber type distribution.
Poultry science (2022-10-12)
Huilin Cheng, Sumin Song, Tae Sub Park, Gap-Don Kim
ABSTRACT
The proteolysis trends and meat quality of the chicken pectoralis major (PM) and iliotibialis (IL) muscles stored at 4°C for 7 d were investigated. After 7 d of storage, the purge loss was higher (P < 0.05) in PM than in IL muscle. The difference in the composition of muscle fibers between PM (100% fast type) and IL (88.85% fast and 11.15% slow types) resulted in differences in proteolysis. Fructose-bisphosphate aldolase, troponin I, myosin heavy chain, and malate dehydrogenase exhibited the same tendencies, but pyruvate kinase, creatine kinase, L-lactate dehydrogenase, and triosephosphate isomerase exhibited different tendencies in the 2 muscles. The activity of cathepsin B was higher in PM than in IL during storage (P < 0.05). These results indicate that the proteolysis trend and changes in meat quality during cold storage are dependent on the different muscle fiber characteristics.
Sigma-Aldrich
Cathepsin Inhibitor I, The Cathepsin Inhibitor I controls the biological activity of Cathepsin. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.