MilliporeSigma
  • Loss of Bcl-3 delays bone fracture healing through activating NF-κB signaling in mesenchymal stem cells.
Loss of Bcl-3 delays bone fracture healing through activating NF-κB signaling in mesenchymal stem cells.
Journal of orthopaedic translation (2022-10-04)
Fuxiao Wang, Jiawei Guo, Yili Wang, Yan Hu, Hao Zhang, Jiao Chen, Yingying Jing, Liehu Cao, Xiao Chen, Jiacan Su
ABSTRACT
Bone fracture healing is a postnatal regenerative process in which fibrocartilaginous callus formation and bony callus formation are important. Bony callus formation requires osteoblastic differentiation of MSCs. The formation of callus was assessed by μCT, Safranin-O, H&E and Masson trichrome staining. Osteogenesis of MSCs was analyzed by ALP staining, ARS staining, qRT-PCR and WB. And we also used IF and TOP/FOP Flash luciferase reporter to assess the nuclear translocation of PP65. In this study, we found Bcl-3 showed a significant correlation with bone fracture healing. Results of μCT showed that loss of Bcl-3 delays bone fracture healing. Safranin-O, H&E and Masson trichrome staining confirmed that loss of Bcl-3 impacted the formation of cartilage and woven bone in callus. Further experiments in vitro manifested that Bcl-3-knockdown could inhibit MSCs osteoblastic differentiation through releasing the inhibition on NF-κB signaling by Co-IP, IF staining and luciferase reporter assay. We unveiled that loss of Bcl-3 could lead to inhibited osteogenic differentiation of MSCs via promoting PP65 nuclear translocation. Our data demonstrated that overexpression of Bcl-3 accelerates bone fracture healing, which serves as a promising therapeutic target for bone fracture treatment.
Product Description
rProtein A Sepharose Fast Flow, Cytiva 17-1279-01, pack of 5 mL