MilliporeSigma
  • Evaluation of glycofurol-based gel as a new vehicle for topical application of naproxen.
Evaluation of glycofurol-based gel as a new vehicle for topical application of naproxen.
AAPS PharmSciTech (2010-07-24)
Nahla Salah Barakat
ABSTRACT
In view of the good skin tolerability, glycofurol was used as a vehicle-based gel, and its effect in the topical penetration of Naproxen (NAP) was investigated. The aims of this study were to develop a suitable gel with bioadhesive property, spreadability, and viscosity for topical anti-inflammatory effect. Three gelling and adhesive agents were examined: Carbopol 974P, Gantrez AN 119, and polyvinylpyrollidone K30. Skin permeation rates and lag times of NAP were evaluated using the Franz-type diffusion cell in order to optimize the gel formulation. The permeation rate of NAP-based gel across the excised rat skin was investigated. A significant increase in permeability parameters such as steady-state flux (J(ss)), permeability coefficient (K(p)), and penetration index (PI) was observed in optimized formulation containing 2% Transcutol as an permeation enhancer. From skin irritation test, it was concluded that the optimized novel glycofurol-based gel formulation was safe to be used for topical drug delivery. The developed glycofurol-based gel appeared promising for dermal and transdermal delivery of naproxen and could be applicable with water-insoluble drugs, which would circumvent most of the problems associated with drug therapy.
Sigma-Aldrich
Tetraglycol, BioXtra