MilliporeSigma
  • Laser-patterned paper-based flow-through filters and lateral flow immunoassays to enable the detection of C-reactive protein.
Laser-patterned paper-based flow-through filters and lateral flow immunoassays to enable the detection of C-reactive protein.
Talanta (2021-11-22)
P P Galanis, I N Katis, P J W He, A H Iles, A J U Kumar, R W Eason, C L Sones
ABSTRACT
We report the use of a laser-based fabrication process in the creation of paper-based flow-through filters that when combined with a traditional lateral flow immunoassay provide an alternative pathway for the detection of a pre-determined analyte over a wide concentration range. The laser-patterned approach was used to create polymeric structures that alter the porosity of the paper to produce porous flow-through filters, with controllable levels of porosity. When located on the top of the front end of a lateral flow immunoassay the flow-through filters were shown to block particles (of known sizes of 200 nm, 500 nm, 1000 nm and 3000 nm) that exceed the effective pore size of the filter while allowing smaller particles to flow through onto a lateral flow immunoassay. The analyte detection is based on the use of a size-exclusive filter that retains a complex (∼3 μm in size) formed by the binding of the target analyte with two antibodies each of which is tagged with different-sized labels (40 nm Au-nanoparticles and 3 μm latex beads), and which is larger than the effective pore size of the filter. This method was tested for the detection of C-reactive protein in a broad concentration range from 10 ng/ml to 100,000 ng/ml with a limit-of-detection found at 13 ng/ml and unlike other reported methods used for analyte detection, with this technique we are able to counter the Hook effect which is a limiting factor in many lateral flow immunoassays.
Sigma-Aldrich
C Reactive Protein human, recombinant, expressed in E. coli, liquid