MilliporeSigma
  • Alzheimer's disease protease-containing plasma extracellular vesicles transfer to the hippocampus via the choroid plexus.
Alzheimer's disease protease-containing plasma extracellular vesicles transfer to the hippocampus via the choroid plexus.
EBioMedicine (2022-02-28)
Jung-Hyun Lee, Christian Ostalecki, Timo Oberstein, Stefan Schierer, Elisabeth Zinser, Martin Eberhardt, Katja Blume, Bianca Plosnita, Lena Stich, Heiko Bruns, Roland Coras, Julio Vera-Gonzales, Manuel Maler, Andreas S Baur
ABSTRACT
Plasma extracellular vesicles (pEV) can harbor a diverse array of factors including active proteases and the amyloid-precursor-protein (APP) cleavage product Aβ, involved in plaque formation in Alzheimer`s diseases (AD). A potential role of such vesicles in AD pathology is unexplored. In a case-control study of randomly selected patients with AD and other neurological diseases (n = 14), and healthy controls (n = 7), we systematically analyzed the content of pEV, using different assay systems. In addition, we determined their entry path into brain tissue, employing animal (mice) injection experiments with ex vivo generated EV that were similar to AD-pEV, followed by multi antigen analysis (MAA) of brain tissue (n = 4 per condition). The results were compared with an IHC staining of human brain tissue in a small cohort of AD patients (n = 3) and controls with no neurodegenerative diseases (n = 3). We show that pEV levels are considerably upregulated in AD patients. Besides numerous inflammatory effectors, AD-pEV contained α-, β- and γ-secretases, able to cleave APP in in target cells. In vitro generated EV with similar characteristics as AD-pEV accumulated in the choroid plexus (CP) of injected animals and reached primarily hippocampal neurons. Corroborating findings were made in human brain samples. An inhibitor of hyaluronic-acid-synthetase (HAS) blocked uploading of proteases and Hyaluronan onto EV in vitro and abolished CP targeting in animal injection experiments. We conclude that protease-containing pEV could be part of a communication axis between the periphery and the brain that could be become detrimental depending on pEV concentration and duration of target cell impact. See the Acknowledgements section.
Sigma-Aldrich
Anti-Alzheimer Precursor Protein A4 Antibody, clone 22C11, Alexa488 Conj., clone 22C11, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor488 conjugated, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-β-Tubulin−FITC antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture