MilliporeSigma
  • Magnetic mixed hemimicelles dispersive solid-phase extraction based on ionic liquid-coated attapulgite/polyaniline-polypyrrole/Fe3O4 nanocomposites for determination of acaricides in fruit juice prior to high-performance liquid chromatography-diode array detection.
Magnetic mixed hemimicelles dispersive solid-phase extraction based on ionic liquid-coated attapulgite/polyaniline-polypyrrole/Fe3O4 nanocomposites for determination of acaricides in fruit juice prior to high-performance liquid chromatography-diode array detection.
Talanta (2017-02-19)
Xiaoling Yang, Kexin Qiao, Fang Liu, Xiaoling Wu, Miyi Yang, Jing Li, Haixiang Gao, Sanbing Zhang, Wenfeng Zhou, Runhua Lu
ABSTRACT
In this research, a novel method using magnetic mixed hemimicelles dispersive solid-phase extraction (MMHDSPE) based on C16mimBr-coated attapulgite/polyaniline-polypyrrole/Fe3O4 (ATP/PANI-PPY/Fe3O4) nanocomposites were investigated for enrichment and separation of three acaricides in fruit juice. In this method, we combined the simplicity and speed of dispersive solid-phase extraction, the advantages of mixed hemimicelles and the facility of the phase separation of the magnetic nanoparticles to develop a simple, rapid, sensitive, and effective method for detecting target analytes from the juice samples. ATP/PANI-PPY/Fe3O4 nanocomposites were successfully synthesized using a one-pot method. The as-prepared nanocomposite sorbents were characterized by Fourier transform infrared spectra (FT-IR), X-ray diffraction (XRD), scanning electron microscopy (SEM), and transmission electron microscopy (TEM). The experimental factors affecting the extraction efficiency including the sorbent type, amount of ATP/PANI-PPY/Fe3O4 nanocomposites, volume of ILs, sonication time, pH, and desorption conditions were optimized. Under the optimal conditions, good linearity was observed for all target analytes, with correlation coefficients (r2) ranging from 0.9994 to 0.9999; the limits of detection (LOD) were in the range of 0.16-0.57μgL-1, and the recoveries of analytes using the proposed method ranged between 88.67% and 95.10%. The sorbents exhibited excellent repeatability in the range of 1.83-4.24% in extracting the three target analytes. In addition, the intra-day and inter-day precision values were found to be in the range of 0.19-6.24% and 2.23-8.36%, respectively. The method was successfully applied to analyze fruit juice samples by rapid preconcentration of acaricides.
Product Description
Supelco
Clofentezine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland