MilliporeSigma
Comparative pathology of murine mucolipidosis types II and IIIC.
Veterinary pathology (2009-03-06)
P Vogel, B J Payne, R Read, W-S Lee, C M Gelfman, S Kornfeld
ABSTRACT
UDP-GlcNAc: lysosomal enzyme N-acetylglucosamine-1-phosphotransferase (GlcNAc-1-phosphotransferase) is an alpha(2)beta(2)gamma(2) hexameric enzyme that catalyzes the first step in the synthesis of the mannose 6-phosphate targeting signal on lysosomal hydrolases. In humans, mutations in the gene encoding the alpha/beta subunit precursor give rise to mucolipidosis II (MLII), whereas mutations in the gene encoding the gamma subunit cause the less severe mucolipidosis IIIC (MLIIIC). In this study we describe the phenotypic, histologic, and serum lysosomal enzyme abnormalities in knockout mice lacking the gamma subunit and compare these findings to those of mice lacking the alpha/beta subunits and humans with MLII and MLIIIC. We found that both lines of mutant mice had elevated levels of serum lysosomal enzymes and cytoplasmic alterations in secretory cells of several exocrine glands; however, lesions in gamma-subunit deficient (Gnptg(-/-)) mice were milder and more restricted in distribution than in alpha/beta-subunit deficient (Gnptab(-/-)) mice. We found that onset, extent, and severity of lesions that developed in these two different knockouts correlated with measured lysosomal enzyme activity; with a more rapid, widespread, and severe storage disease phenotype developing in Gnptab(-/-) mice. In contrast to mice deficient in the alpha/beta subunits, the mice lacking the gamma subunits were of normal size, lacked cartilage defects, and did not develop retinal degeneration. The milder disease in the gamma-subunit deficient mice correlated with residual synthesis of the mannose 6-phosphate recognition marker. Of significance, neither strain of mutant mice developed cytoplasmic vacuolar inclusions in fibrocytes or mesenchymal cells (I-cells), the characteristic lesion associated with the prominent skeletal and connective tissue abnormalities in humans with MLII and MLIII. Instead, the predominant lesions in both lines of mice were found in the secretory epithelial cells of several exocrine glands, including the pancreas, and the parotid, submandibular salivary, nasal, lacrimal, bulbourethral, and gastric glands. The absence of retinal and chondrocyte lesions in Gnptg(-/-) mice might be attributed to residual beta-glucuronidase activity. We conclude that mice lacking either alpha/beta or gamma subunits displayed clinical and pathologic features that differed substantially from those reported in humans having mutations in orthologous genes.
Sigma-Aldrich
4-Methylumbelliferyl α-D-mannopyranoside, ≥97% (HPLC)