MilliporeSigma
  • Prognostic value of the PrPC-ILK-IDO1 axis in the mesenchymal colorectal cancer subtype.
Prognostic value of the PrPC-ILK-IDO1 axis in the mesenchymal colorectal cancer subtype.
Oncoimmunology (2021-07-13)
Alexandre Ghazi, Delphine Le Corre, Camilla Pilati, Julien Taieb, Thomas Aparicio, Audrey Didelot, Shoukat Dedhar, Claire Mulot, Karine Le Malicot, Fatima Djouadi, Aurélien de Reynies, Jean-Marie Launay, Pierre Laurent-Puig, Sophie Mouillet-Richard
ABSTRACT
The CMS4 mesenchymal subtype of colorectal cancer (CRC) is associated with poor prognosis and resistance to treatment. The cellular prion protein PrPC is overexpressed in CMS4 tumors and controls the expression of a panel of CMS4-specific genes in CRC cell lines. Here, we sought to investigate PrPC downstream pathways that may underlie its role in CMS4 CRC. By combining gene set enrichment analyses and gain and loss of function approaches in CRC cell lines, we identify the integrin-linked kinase ILK as a proximal effector of PrPC that mediates its control on the CMS4 phenotype. We further leveraged three independent large CRC cohorts to assess correlations in gene expression pattern with patient outcomes and found that ILK is overexpressed in CMS4 mesenchymal tumors and confers a poor prognosis, especially when combined with high expression of the PrPC encoding gene PRNP. Of note, we discovered that the PrPC-ILK signaling axis controls the expression and activity of the tryptophan metabolizing enzyme indoleamine 2,3 dioxygenase IDO1, a key player in immune tolerance. In addition, we monitored alterations in the levels of tryptophan and its metabolites of the kynurenine pathway in the plasma of metastatic CRC patients (n = 325) and we highlight their prognostic value in combination with plasma PrPC levels. Thus, the PrPC-ILK-IDO1 axis plays a key role in the mesenchymal subtype of CRC. PrPC and IDO1-targeted strategies may represent new avenues for patient stratification and treatment in CRC.
Sigma-Aldrich
QLT0267, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid