MilliporeSigma
  • Intermitochondrial signaling regulates the uniform distribution of stationary mitochondria in axons.
Intermitochondrial signaling regulates the uniform distribution of stationary mitochondria in axons.
Molecular and cellular neurosciences (2022-02-09)
Nozomu Matsumoto, Ikuma Hori, Masashi K Kajita, Tomoya Murase, Wataru Nakamura, Takahiro Tsuji, Seiji Miyake, Masaru Inatani, Yoshiyuki Konishi
ABSTRACT
In the central nervous system (CNS), many neurons develop axonal arbors that are crucial for information processing. Previous studies have demonstrated that premature axons contain motile and stationary mitochondria, and their balance is important for axonal arborization. However, the mechanisms by which neurons determine the positions of stationary mitochondria as well as their turnover remain to be elucidated. We observed that the distribution of stationary mitochondrial spots along the unmyelinated and nonsynaptic axons is not random but rather relatively uniform both in primary cultured neurons and in tissues. Intriguingly, whereas the positions of each mitochondrial spot changed over time, the overall distribution remained uniform. In addition, local inactivation of mitochondria by KillerRed mediated chromophore-assisted light inactivation (CALI) inhibited the translocation of mitochondrial spots in adjacent axonal regions, suggesting that functional mitochondria enhance the motility of other mitochondria in the vicinity. Signals of ATP:ADP sensor, PercevalHR indicated that the ATP:ADP ratio was relatively high around mitochondria, and treating axons with phosphocreatine (PCr), which supplies ATP, reduced the immobile mitochondria induced by the local mitochondrial inactivation. In a mathematical model, we found that the ATP gradient generated by mitochondria, and ATP dependent regulation of mitochondrial motility could establish uniform mitochondrial distribution. These observations suggest that axons in the CNS possess the system that distributes mitochondria uniformly, and intermitochondrial signaling contribute to the regulation. In addition, our results suggest the possibility that ATP might be one of the molecules mediating the signaling.
Product Description
SAFC
Basal Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture