MilliporeSigma
  • Treacle and TOPBP1 control replication stress response in the nucleolus.
Treacle and TOPBP1 control replication stress response in the nucleolus.
The Journal of cell biology (2021-06-09)
Artem K Velichko, Natalia Ovsyannikova, Nadezhda V Petrova, Artem V Luzhin, Maria Vorobjeva, Alexey S Gavrikov, Alexander S Mishin, Igor I Kireev, Sergey V Razin, Omar L Kantidze
ABSTRACT
Replication stress is one of the main sources of genome instability. Although the replication stress response in eukaryotic cells has been extensively studied, almost nothing is known about the replication stress response in nucleoli. Here, we demonstrate that initial replication stress-response factors, such as RPA, TOPBP1, and ATR, are recruited inside the nucleolus in response to drug-induced replication stress. The role of TOPBP1 goes beyond the typical replication stress response; it interacts with the low-complexity nucleolar protein Treacle (also referred to as TCOF1) and forms large Treacle-TOPBP1 foci inside the nucleolus. In response to replication stress, Treacle and TOPBP1 facilitate ATR signaling at stalled replication forks, reinforce ATR-mediated checkpoint activation inside the nucleolus, and promote the recruitment of downstream replication stress response proteins inside the nucleolus without forming nucleolar caps. Characterization of the Treacle-TOPBP1 interaction mode leads us to propose that these factors can form a molecular platform for efficient stress response in the nucleolus.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-TCOF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution