MilliporeSigma
  • Disruption of GABAergic tone in the dorsomedial hypothalamus attenuates responses in a subset of serotonergic neurons in the dorsal raphe nucleus following lactate-induced panic.
Disruption of GABAergic tone in the dorsomedial hypothalamus attenuates responses in a subset of serotonergic neurons in the dorsal raphe nucleus following lactate-induced panic.
Journal of psychopharmacology (Oxford, England) (2008-03-01)
Pl Johnson, Ca Lowry, W Truitt, A Shekhar
ABSTRACT
Panic patients are vulnerable to induction of panic attacks by sub-threshold interoceptive stimuli such as intravenous (i.v.) sodium lactate infusions. Facilitation of serotonergic signaling with selective serotonin reuptake inhibitors can suppress anxiety and panic-like responses, but the mechanisms involved are not clearly defined. We investigated the effects of i.v. 0.5 M sodium lactate or saline, in control and panic-prone rats on c-Fos expression in serotonergic neurons within subdivisions of the midbrain/pontine raphe nuclei. Rats were chronically infused with either the GABA synthesis inhibitor l-allylglycine into the dorsomedial hypo thalamus to make them panic-prone, or the enantiomer d-allylglycine (d-AG) in controls. Lactate increased c-Fos expression in serotonergic neurons located in the ventrolateral part of the dorsal raphe nucleus (DRVL) and ventrolateral periaqueductal gray (VLPAG) of control, but not panic-prone, rats. The distribution of lactate-sensitive serotonergic neurons in d-AG-treated rats is virtually identical to previously defined pre-sympathomotor serotonergic neurons with multisynaptic projections to peripheral organs mediating 'fight-or-flight'-related autonomic and motor responses. We hypothesized that serotonergic neurons within the DRVL/VLPAG region represent a 'sympathomotor control system' that normally limits autonomic/behavioral responses to innocuous interoceptive and exteroceptive stimuli, and that dysfunction of this serotonergic system contributes to an anxiety-like state and increases vulnerability to panic in animals and humans.