MilliporeSigma
  • Stability-Indicating HPLC Method for the Determination of Cefcapene Pivoxil.
Stability-Indicating HPLC Method for the Determination of Cefcapene Pivoxil.
Chromatographia (2013-04-05)
Przemysław Zalewski, Judyta Cielecka-Piontek, Piotr Garbacki, Anna Jelińska, Marta Karaźniewicz-Łada
ABSTRACT
The stability-indicating LC assay method was developed and validated for quantitative determination of cefcapene pivoxil in the presence of degradation products formed during forced degradation studies. An isocratic RP-HPLC method was developed with a Lichrospher RP-18 (250 mm × 4.6 mm, 5 μm) column and the mobile phase composed of 45 volumes of acetonitrile and 55 volumes of mixture composed of citric acid 10 mmol L-1 and potassium chloride 18 mmol L-1. The flow rate of the mobile phase was 1 mL min-1. Detection wavelength was 270 nm and temperature was 30 °C. Cefcapene pivoxil, similar to other cephalosporins, was subjected to stress conditions of degradation in aqueous solutions including hydrolysis, oxidation, and thermal degradation. The method was validated with regard to linearity, accuracy, precision, selectivity, and robustness. The method was applied successfully for the determination of cefcapene pivoxil during kinetic studies in aqueous solutions (pH and thermal degradation) and in solid state (oxidative, thermal, and radiolytic degradation).
Sigma-Aldrich
Cefcapene Pivoxil Hydrochloride Hydrate, ≥98% (HPLC)