MilliporeSigma
  • Diallyl disulfide potentiates anti-obesity effect of green tea in high-fat/high-sucrose diet-induced obesity.
Diallyl disulfide potentiates anti-obesity effect of green tea in high-fat/high-sucrose diet-induced obesity.
The Journal of nutritional biochemistry (2018-12-06)
Jaehoon Bae, Motofumi Kumazoe, Yoshinori Fujimura, Hirofumi Tachibana
ABSTRACT
Obesity is a major problem in developed countries and a burden on social health care systems. Several epidemiological studies showed the protective effects of green tea against obesity-related diseases. Cyclic guanosine monophosphate (cGMP) acts as a mediator for the physiological effects of (-)-epigallocatechin-3-O-gallate, the major constituent of green tea. Here, we showed that the level of phosphodiesterase 5, a negative regulator of cGMP, was up-regulated in adipose tissues of high-fat/high-sucrose (HF/HS) diet-fed mice and that this up-regulation was ameliorated by diallyl disulfide (DADS), the major organosulfur in garlic. A green tea extract (GT) and DADS in combination attenuated HF/HS diet-induced adipose increase and triglyceride accumulation in the liver. In these mechanisms, the combination regimen suppressed the HF/HS diet-induced up-regulation of fatty acid synthesis-related enzymes including sterol regulatory element-binding protein-1 (SREBP-1), fatty acid synthase, and stearoyl-CoA desaturase-1. Moreover, this combination diet up-regulated thermogenesis-related genes including peroxisome proliferator-activated receptor (PPAR) gamma coactivator 1 alpha and uncoupling proteins in both white and brown adipose tissues. In conclusion, we identified DADS as an enhancer of the anti-obesity effect of GT accompanied by the suppression of SREBP-1 and activation of PPAR axis. The combination diet is a novel and easily applicable approach against obesity-related diseases.
Sigma-Aldrich
Diallyl disulfide, ≥98% (HPLC)
Pricing and availability is not currently available.