MilliporeSigma
  • Natural autoantibodies to the gonadotropin-releasing hormone receptor in polycystic ovarian syndrome.
Natural autoantibodies to the gonadotropin-releasing hormone receptor in polycystic ovarian syndrome.
PloS one (2021-04-03)
Lisa-Marie Sattler, Hanna A Schniewind, Waldemar B Minich, Christoph W Haudum, Petra Niklowitz, Julia Münzker, Gábor L Kovács, Thomas Reinehr, Barbara Obermayer-Pietsch, Lutz Schomburg
ABSTRACT
Polycystic ovarian syndrome (PCOS) is a complex disease with different subtypes and unclear etiology. Among the frequent comorbidities are autoimmune diseases, suggesting that autoantibodies (aAb) may be involved in PCOS pathogenesis. As the gonadal axis often is dysregulated, we tested the hypothesis that aAb to the gonadotropin-releasing hormone receptor (GnRH-R) are of diagnostic value in PCOS. An in vitro assay for quantifying aAb to the GnRH-R (GnRH-R-aAb) was established by using a recombinant fusion protein of full-length human GnRH-R and firefly luciferase. A commercial rabbit antiserum to human GnRH-R was used for standardization. Serum samples of control subjects and different cohorts of European PCOS patients (n = 1051) were analyzed. The novel GnRH-R-aAb assay was sensitive, and signals were linear on dilution when tested with the commercial GnRH-R antiserum. Natural GnRH-R-aAb were detected in one control (0.25%) and two PCOS samples (0.31%), and 12 samples were slightly above the threshold of positivity. The identification of samples with positive GnRH-R-aAb was reproducible and the signals showed no matrix interferences. Natural GnRH-R-aAb are present in a very small fraction of adult control and PCOS subjects of European decent. Our results do not support the hypothesis that the GnRH-R constitutes a relevant autoantigen in PCOS.
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Protein A Sepharose Cl-4B, Cytiva 17-0963-03, pack of 25 mL