MilliporeSigma
  • Characterisation of a New Molecule Based on Two E2 Sequences from Bovine Viral Diarrhoea-mucosal Disease Virus Fused To the Human Immunoglobulin Fc Fragment.
Characterisation of a New Molecule Based on Two E2 Sequences from Bovine Viral Diarrhoea-mucosal Disease Virus Fused To the Human Immunoglobulin Fc Fragment.
Journal of veterinary research (2021-04-06)
Alaín González Pose, Raquel Montesino Seguí, Rafael Maura Pérez, Florence Hugues Salazar, Ignacio Cabezas Ávila, Claudia Altamirano Gómez, Oliberto Sánchez Ramos, Jorge Roberto Toledo
ABSTRACT
Proper conformational arrangement of the E2 molecules of bovine viral diarrhoea-mucosal disease virus (BVD-MDV) is crucial to obtain an effective recombinant vaccine candidate against the disease. In this study, we characterised a new molecule composed of two distinct sequences of the E2 glycoprotein of BVD-MDV and the Fc fragment of human immunoglobulin (BVDE2Fc). The chimaeric protein was expressed in mammalian cell lines of different species by adenoviral transduction and purified by immobilised metal-affinity chromatography. The N-glycans were profiled by HPLC, and the BVDE2Fc immunogenicity was assessed in male mice. The antigen-antibody reactions were evaluated by ELISA. The MDBK cell line was selected from among five for the final production of BVDE2Fc. After purification to over 90%, the N-glycan profile showed neutral and complex oligosaccharides. The mouse immunisation induced a strong humoral response, which produced antibodies able to attach to conformational epitopes on E2 molecules, while the Fc fragment barely contributed to the immune response. Additionally, BVDE2Fc attached to antibodies from bovine sera positive to distinct BVD-MDV subtypes, whereas the loss of BVDE2Fc structure during the deglycosylation process considerably diminished those interactions. These results demonstrate that the structure of E2 molecules arranged in tandem and attached to an Fc fragment could represent a viable design for future vaccine candidates against BVD-MD.
Sigma-Aldrich
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder